Learn More
Medchemexpress LLC HER4 Rat HEK293 H 50ug
SDP
Catalog No. 5000180472
Encompass_Preferred
HER4 Protein crucial in central nervous system development influences neural crest cell migration and axon guidance Additionally it participates in mammary gland differentiation inducing milk proteins and contributing to lactation HER4 Protein Rat (HEK293 His) is the recombinant rat-derived HER4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80472 Supplier Medchemexpress LLC Supplier No. HYP7303750UG
HER4 Protein crucial in central nervous system development influences neural crest cell migration and axon guidance Additionally it participates in mammary gland differentiation inducing milk proteins and contributing to lactation HER4 Protein Rat (HEK293 His) is the recombinant rat-derived HER4 protein expressed by HEK293 with C-His labeled tag