Learn More
Thermo Scientific™ O2 Control
Catalog No. 13642691 Shop All Thermo Scientific Products
Change view
Click to view available options
Oxygen Control:
1 to 21%
Catalog No. Oxygen Control
13-642-691 1 to 21%
1 options
Catalog No. 13-642-691 Supplier Thermo Scientific™ Supplier No. 51901137
Add to Cart
Add to Cart
For Use With: Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
Upgrade your incubators to take advantage of O2 control.
TRUSTED_SUSTAINABILITY
Specifications
Description 1 to 21% O2 control
For Use With (Equipment) Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
Lighting LED
Oxygen Control 1 to 21%
Type O2 Control