Description
Material: Glass; For Use With: Heracell VIOS 250i CO2 Incubator
- O2 control
- Gas tight screen
- 6 inner glass doors
- 1/2 width shelves
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | O2 1-21% Control Glass Doors |
| For Use With (Equipment) | Heracell VIOS 250i CO2 Incubator |
| Lighting | LED |
| Material | Glass |
| Oxygen Control | 1 to 21% |
| Type | O2 Control Doors |
| Gas Type | O2 |
| Includes | O2 control with gas tight screen, 6 inner glass doors,1/2 width shelves |