Learn More
Medchemexpress LLC HY-N4053 5mg Medchemexpress, Heraclenin CAS:2880-49-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN40535MG
Encompass_Preferred
Medchemexpress, HY-N4053 5mg Heraclenin CAS:2880-49-1 Heraclenin, a natural furanocoumarin, isolated from the root parts of the plant Prangos pabularia. Heraclenin significantly inhibits T cell receptor-mediated proliferation in human primary T cells in a concentration-dependent manner by targeting nuclear factor of activated T-cells (NFAT). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2919