Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Floor Stands
GSA_VA
Catalog No. 13998008
Change view
Click to view available options
Product Type:
Adjustable Floor Stand
Castors
Electronic Adjustable Floor Stand
For Use With (Equipment):
Heraguard ECO 0.9
Heraguard ECO 0.9, 1.2, 1.5, 1.8
Heraguard ECO 1.2
Heraguard ECO 1.5
Heraguard ECO 1.8, 1.8/95
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
13-998-008 Adjustable Floor Stand Heraguard ECO 1.5
01-257-739 Electronic Adjustable Floor Stand Heraguard ECO 1.5
01-257-740 Electronic Adjustable Floor Stand Heraguard ECO 0.9
13-998-009 Adjustable Floor Stand Heraguard ECO 1.8, 1.8/95
13-998-007 Adjustable Floor Stand Heraguard ECO 1.2
13-998-032 Castors Heraguard ECO 0.9, 1.2, 1.5, 1.8
01-257-742 Electronic Adjustable Floor Stand Heraguard ECO 1.2
01-257-741 Electronic Adjustable Floor Stand Heraguard ECO 1.8, 1.8/95
13-998-006 Adjustable Floor Stand Heraguard ECO 0.9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-998-008 Supplier Thermo Scientific™ Supplier No. 50116444
Only null left
Add to Cart
Add to Cart
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Choose from fixed, manual and electrically adjustable floor stands for the Thermo Scientific™ Heraguard™ ECO clean bench.
Choose from fixed, manual and electrically adjustable Thermo Scientific™ Heraguard™ ECO Clean Bench Floors Stands for the Heraguard™ ECO clean bench.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Heraguard ECO 1.5
Description Provides variable ECO work height of 760 mm - 960 mm in 50 mm adjustable increments.
Product Type Adjustable Floor Stand
Height Overall (Metric) 680 to 880 mm