Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heratherm CO2 Chamber 750 L

Catalog No. 13642652 Shop All Thermo Scientific Products
Change view
Click to view available options
Door Style:
Glass door
Solid (outer), Glass (inner)
Solid outer door with inner glass door
Capacity (Metric):
247 L
750 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style Capacity (Metric)
13-642-652 Solid outer door with inner glass door 247 L
01-672-444 Solid (outer), Glass (inner) 750 L
01-672-445 Glass door 750 L
13-642-651 Glass door 247 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-642-652 Supplier Thermo Scientific™ Supplier No. ECS250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Precisely control the essential parameters of your distinct cell growth or CO2 testing conditions — active humidity controls the incubation environment with accuracy.

Accurate control of three parameters - Temperature above and below ambient, relative humidity and CO2 - provide great environment in Cell growth and testing applications for reproducible results.

  • Uniform Temperature / CO2 / relative humidity throughout the chamber
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • CO2 control system ensures stable CO2 environment, range of 0 – 20 Vol-%.
  • Environmentally friendly water-efficient humidification system - with water tank or optional fixed water connection for in-house purified water
  • Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option.
Choose from a variety of country-specific sockets. (Ordered with separate order code)

TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 8.8 cu.ft.
Capacity (English) Shelf 66.1 lb.
Capacity (Metric) Shelf 30 kg
Certifications/Compliance UL, CE
Data Outputs USB, Thermo Cloud via OPC UA
Capacity (Metric) 247 L
Chamber Material Powder-coated Cold Rolled Steel
Lighting No
Material Powder-coated Cold Rolled Steel
No. of Shelves 2
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Temperature Range (Metric) 0 to 70°C (15 to 45°C with CO2)
Type Growth Chamber
Display 7 in. Touchscreen user interface
Door Style Solid outer door with inner glass door
For Use With (Application) Cell growth, testing
Humidity Range 10 to 90% RH
Humidity Source Removable water tank
No. of Casters None
Ports 2 access ports
Programmability Temperature and humidity ramping, CO2
Shelf Area (English) 3.05 ft.2
Shelf Area (Metric) 0.28 m2
Shelf Material Stainless Steel (304/1.4301)
Temperature Accuracy Microprocessor - programmable
Temperature Resolution 0.1°C
Temperature Stability ≤ ±0.1°C @ 20°C to 37°C
Temperature Uniformity ≤ ±0.5°C @ 20°C to 37°C
Warranty 24 months
Water Consumption ≤ 0.25 L per day - Value depends on settings, usage (e.g. number of door openings), and environmental conditions
Water Resistivity 0.05 - 1 MΩ
Width (English) 35.5 in.
Width (English) Interior 26 in.
Width (English) Shelf 24.2 in.
Width (Metric) 901 mm
Width (Metric) Interior 660 mm
Width (Metric) Shelf 615 mm
Height (English) 44.4 in.
Height (English) Interior 31.5 in.
Height (Metric) 1128 mm
Height (Metric) Interior 800 mm
Depth (English) 31.1 in.
Depth (English) Interior 20.4 in.
Depth (Metric) 790 mm
Depth (Metric) Interior 519 mm
Shipping Weight (English) 459 lb.
Shipping Weight (Metric) 208 kg
Electrical Requirements 100/240 V, 50/60 Hz
Voltage 100/115/220/230 V
Wattage 900 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.