Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heratherm Incubation Chamber PROMO

Catalog No. 13642653PM
Change view
Click to view available options
Door Style:
Glass Door
Solid Outer Door, Glass Inner Door
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style
13-642-653PM Glass Door
13-642-654PM Solid Outer Door, Glass Inner Door
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-642-653PM Supplier Thermo Scientific™ Supplier No. EIG250PM
Get Up to 40% Off Equipment
View all promotions
Add to Cart
Add to Cart

Comprehensive data monitoring and temperature control combined with a flexible Heratherm Incubation Chamber provide a dependable option for microbiological incubation or temperature-dependent storage and testing.

  • Uniform Temperature throughout the chamber.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 8.8 cu.ft.
Capacity (English) Shelf 66.1 lb.
Capacity (Metric) Shelf 30 kg
Certifications/Compliance UL, CE
Data Outputs USB, Thermo Cloud via OPC UA
Capacity (Metric) 247 L
Chamber Material Powder-coated Cold Rolled Steel
Lighting No
Material Powder-coated Cold Rolled Steel
No. of Shelves 2
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Temperature Range (Metric) 0°C to 70°C
Type Temperature Test Chamber
Display 7 in. Touchscreen
Door Style Glass Door
For Use With (Application) Incubation, Storage, Temperature Testing
No. of Casters None
Ports 2
Programmability Temperature Ramping
Shelf Area (English) 3.05 ft.2
Shelf Area (Metric) 0.28 m2
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor - Programmable
Temperature Resolution 0.1°C
Temperature Stability ≤ ±0.1°C at 20°C to 37°C
Temperature Uniformity ≤ ±0.5°C at 20°C to 37°C
Warranty 24 Months
Width (English) 35.5 in.
Width (English) Interior 26 in.
Width (English) Shelf 24.2 in.
Width (Metric) 901 mm
Width (Metric) Interior 660 mm
Width (Metric) Shelf 615 mm
Height (English) 44.4 in.
Height (English) Interior 31.5 in.
Height (Metric) 1128 mm
Height (Metric) Interior 800 mm
Depth (English) 31.1 in.
Depth (English) Interior 20.4 in.
Depth (Metric) 790 mm
Depth (Metric) Interior 519 mm
Shipping Weight (English) 448 lb.
Shipping Weight (Metric) 203 kg
Electrical Requirements 100/240V, 50/60 Hz
Voltage 100/115/220/230 V
Wattage 600 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.