Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heratherm Stability Chamber

Catalog No. 01672439PM Shop All Thermo Scientific Products
Change view
Click to view available options
Door Style:
Glass
Solid Outer, Glass Inner
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style
01-672-439PM Glass
01-672-438PM Solid Outer, Glass Inner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 01-672-439PM Supplier Thermo Scientific™ Supplier No. ESG750PM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Gain confidence in your product when testing stability and shelf life with robust Heratherm Stability chambers designed to meet a variety of quality control demands.

  • Broad Relative Humidity range to cover many applications, including Humidity levels below ambient.
  • Environmentally friendly water-efficient humidification system.
  • Minimal condensate in standard applications.
  • Integrated water tank; fixed water connection for in-house purified water option available.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
TRUSTED_SUSTAINABILITY

Specifications

Alarm Types Audible and Visible
Capacity (English) 26.5 cu.ft.
Capacity (English) Shelf 66.1 lb.
Capacity (Metric) Shelf 30 kg
Certifications/Compliance UL, CE
Data Outputs USB, Ethernet
Capacity (Metric) 750 L
Chamber Material Powder-coated Cold Rolled Steel
Lighting No
Material Powder-coated Cold Rolled Steel
No. of Shelves 3
Plug Type Nema 5-20P
Relative Humidity 10 to 90%
Temperature Range (Metric) 0 to 70°C
Type Humidity Chamber
Display Touchscreen
Door Style Glass
Humidity Range 10 to 90% RH
Humidity Resolution 0.1% RH
No. of Casters 4
Ports 2 Access Ports
Programmability Temperature and Humidity Ramping
Shelf Area (English) 4.9 ft.2
Shelf Area (Metric) 0.46 m2
Shelf Material Stainless Steel (304/1.4301)
Temperature Control Microprocessor
Temperature Resolution 0.1°C
Temperature Stability ≤ ±0.1°C at 20°C to 37°C
Temperature Uniformity ≤ ±0.5°C at20°C to 37°C
Warranty 24 Months
Water Consumption 0.15 to 1 L/day
Water Resistivity 0.05 to 1 MOhm
Width (English) 42.52 in.
Width (English) Interior 33.07 in.
Width (English) Shelf 22.6 in.
Width (Metric) 108 cm
Width (Metric) Interior 84 cm
Width (Metric) Shelf 79.5 cm
Height (English) 72.24 in.
Height (English) Interior 57.09 in
Height (Metric) 183.5 cm
Height (Metric) Interior 145 cm
Depth (English) 34.84 in.
Depth (English) Interior 24.8 in.
Depth (Metric) 88.5 cm
Depth (Metric) Interior 63 cm
Shipping Weight (English) 820 lb.
Shipping Weight (Metric) 372 kg
Electrical Requirements 100/240 V, 50/60 Hz, 1400 W
Voltage 100/115/220/230 V
Wattage 1400 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.