Learn More
TARGETMOL CHEMICALS INC HERBACETIN 10MG
SDP
Catalog No. 502579713
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Herbacetin induces apoptosis in HepG2 cells by ROS and PI3K/Akt pathway. 2. Herbacetin suppresses the HGF-induced motility of human breast cancer MDA-MB-231 cells by inhibiting c-Met and Akt phosphorylation. purity: 99%
Catalog No. 50-257-9713 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S133110MG
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Herbacetin induces apoptosis in HepG2 cells by ROS and PI3K/Akt pathway. 2. Herbacetin suppresses the HGF-induced motility of human breast cancer MDA-MB-231 cells by inhibiting c-Met and Akt phosphorylation. purity: 99%