Learn More
Cayman Chemical HerbImycIn C 2.5mg
SDP
Catalog No. 503015809
Encompass_Preferred
A bacterial metabolite; cytotoxic to HeLa and Ehrlich cells (IC50s = 7.3 and 1.2 μg/ml, respectively)
Catalog No. 50-301-5809 Supplier Cayman Chemical Supplier No. 254852.5 MG
A bacterial metabolite; cytotoxic to HeLa and Ehrlich cells (IC50s = 7.3 and 1.2 μg/ml, respectively)