Learn More
Aviva Systems Biology Corp HERC4 Antibody - middle region (ARP43298_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43298_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HERC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HERC4. Alternate names: DKFZP564G092, KIAA1593ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-58487