Learn More
Aviva Systems Biology Corp HERC4 Antibody - middle region (ARP42991_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42991_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HERC4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human HERC4. Alternate names: DKFZp564G092, KIAA1593ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-050