Learn More
Aviva Systems Biology Corp HERC5 Antibody - N-terminal region (ARP43212_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43212_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HERC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HERC5. Alternate names: CEB1, CEBP1ApplicationWB. Species reactivity: Cow, Dog, Human
Catalog No. 50-014-58461