Learn More
Agilent Technologies Agilent Herculase II Fusion DNA Polymerase, High Fidelity, High Yield, 40 Rxn
Catalog No. NC3932872
Encompass
This fusion DNA polymerase is engineered for high-performance PCR, delivering superior yields and reliability with shorter cycling times. It is particularly well-suited for routine applications, long-range PCR, and the amplification of complex or GC-rich targets. The formulation includes the ArchaeMaxx polymerase enhancing factor to improve performance and sensitivity, and it is capable of amplifying targets with up to 84% GC content.
  • Superior yield for routine and difficult PCR applications
  • Reduced overall run time with extension times as short as 15 sec/kb
  • High sensitivity for the amplification of low-abundance targets
  • Equivalent fidelity to Pfu DNA polymerase
  • Successful amplification of GC-rich targets up to 84%
  • Optimized for PCR cloning, RT-PCR, site-directed mutagenesis, and NGS library preparation
  • Compatible with common workflow protocols such as SureSelect
Catalog No. NC3932872 Supplier Agilent Technologies Supplier No. 600675
This fusion DNA polymerase is engineered for high-performance PCR, delivering superior yields and reliability with shorter cycling times. It is particularly well-suited for routine applications, long-range PCR, and the amplification of complex or GC-rich targets. The formulation includes the ArchaeMaxx polymerase enhancing factor to improve performance and sensitivity, and it is capable of amplifying targets with up to 84% GC content.
  • Superior yield for routine and difficult PCR applications
  • Reduced overall run time with extension times as short as 15 sec/kb
  • High sensitivity for the amplification of low-abundance targets
  • Equivalent fidelity to Pfu DNA polymerase
  • Successful amplification of GC-rich targets up to 84%
  • Optimized for PCR cloning, RT-PCR, site-directed mutagenesis, and NGS library preparation
  • Compatible with common workflow protocols such as SureSelect