Learn More
Medchemexpress LLC HY-P7365 10ug Medchemexpress, Heregulin-1 β1, Human CAS: Purity:>98%
SDP
Catalog No. 501934383
Encompass_Preferred
Medchemexpress, HY-P7365 10ug Heregulin-1 β1, Human CAS: Recombinant Human Heregulin-1 beta1 belongs to a family of structurally-related polypeptide growth factors derived from alternatively spliced genes, induces Fn14 expression in MCF7 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4383 Supplier Medchemexpress LLC Supplier No. HYP736510UG
Medchemexpress, HY-P7365 10ug Heregulin-1 β1, Human CAS: Recombinant Human Heregulin-1 beta1 belongs to a family of structurally-related polypeptide growth factors derived from alternatively spliced genes, induces Fn14 expression in MCF7 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.