Learn More
Enzo Life Sciences HERG1 (CT) (pan) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX215049R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant HERG1 (human ether-a-go-go related protein 1) (aa 1106-1158). Species reactivity: Human, Mouse. Specificity: Recognizes C-terminal epitope of HERG1 and mouse Erg1. Recognizes both isoforms HERG1A and HERG1B. Applications: Flow Cytometry, IHC (FS), IHC (PS), WB. Formulation: Liquid. Neat serum. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8208
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.