Learn More
Enzo Life Sciences HERG1B polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX215051R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 15-28 (R15PRAQKGRVRRAVR28) of HERG1B (human ether-a-go-go related protein 1B). UniProt ID: Q12809. Species reactivity: Human, Mouse. Specificity: Recognizes HERG1B and mouse Erg1B isoform. Applications: ICC, IHC (FS), WB. Purity: Affinity purified. Formulation: Liquid. In PBS, pH 7.2.  . Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. 50-200-8210