Learn More
TA Instruments Hermetic Lids with Pinhole, Laser Cut Vent Hole, 75 Micron Diameter, Vapor Pressure Studies, Temperature Range -180°C to 600°C, Improve Resolution of Volatile Species, 50 Pack
Catalog No. 5000580678 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Hermetic Lids with Pinhole are special hermetic sample pan lids with a laser-cut vent hole (75 micron diameter) used for vapor pressure studies. Temperature range is -180°C to 600°C. These lids improve resolution for volatile species analysis.
Catalog No. 50-005-80678 Supplier TA Instruments Supplier No. 901698.901
Waters TA Instruments Hermetic Lids with Pinhole are special hermetic sample pan lids with a laser-cut vent hole (75 micron diameter) used for vapor pressure studies. Temperature range is -180°C to 600°C. These lids improve resolution for volatile species analysis.