Learn More
Alkali Scientific Hermle Z167-M Microcentrifuge
SDP
Catalog No. 503029567
Encompass_Preferred
The Hermle Z167-M Microcentrifuge is a powerful compact easy-to-use high speed microcentrifuge for labs looking to save valuable bench space but not compromise on speed and power The Hermle Z167-M is compact design houses a powerful maintenance free induction motor that can reach up to 14000rpm/15994 xg The included rotor is an 18-place Accompanied by an easy to use control panel the unique EZ-Scrolltouch pad gives you ease of control and a modern feel Additionally this model features a color changing LED status indicator that lets you know what operating state the unit is in from a distance Whether the unit is running on standby or in sleep mode it is easy to tell from a far with this new feature The advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf in increments of 10 In addition to retrieving saved programs the easy to use front panel can be used to adjust accel/decel rates run time and change the end of run alarm tone
Catalog No. 50-302-9567 Supplier Alkali Scientific Supplier No. RS6208M
Add to Cart
Add to Cart
The Hermle Z167-M Microcentrifuge is a powerful compact easy-to-use high speed microcentrifuge for labs looking to save valuable bench space but not compromise on speed and power The Hermle Z167-M is compact design houses a powerful maintenance free induction motor that can reach up to 14000rpm/15994 xg The included rotor is an 18-place Accompanied by an easy to use control panel the unique EZ-Scrolltouch pad gives you ease of control and a modern feel Additionally this model features a color changing LED status indicator that lets you know what operating state the unit is in from a distance Whether the unit is running on standby or in sleep mode it is easy to tell from a far with this new feature The advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf in increments of 10 In addition to retrieving saved programs the easy to use front panel can be used to adjust accel/decel rates run time and change the end of run alarm tone