Learn More
Alkali Scientific Hermle Z206-A Compact Centrifu
SDP
Catalog No. 503029639
Encompass_Preferred
The Hermle Z206-A Compact Centrifuge is suitable for a variety of applications inside both the clinical and research setting Three angle rotor options are available permitting use with all common tube sizes from 0 2ml to 50ml A unique 6 x 5ml swing out rotor is also available for clinical applications requiring horizontal separation within 5ml blood collection tubes The advanced microprocessor controls feature an ergonomic turn knob that controls all settings (time speed accel/decel levels etc ) The speed can be adjusted from 200 rpm up to a maximum speed of 6000 rpm (corresponding to 4427 xg) in the popular 50ml and 15ml rotors The centrifuge lid features and automatic lid-lock that prevents it from being opened while the rotor is still in motion In addition a unique imbalance detection system is designed to automatically power down the motor in the event of improper loading of the rotor or any tube breakage Compact quiet and versatile are just a few of the features found in th
Catalog No. 50-302-9639 Supplier Alkali Scientific Supplier No. RS6201
The Hermle Z206-A Compact Centrifuge is suitable for a variety of applications inside both the clinical and research setting Three angle rotor options are available permitting use with all common tube sizes from 0 2ml to 50ml A unique 6 x 5ml swing out rotor is also available for clinical applications requiring horizontal separation within 5ml blood collection tubes The advanced microprocessor controls feature an ergonomic turn knob that controls all settings (time speed accel/decel levels etc ) The speed can be adjusted from 200 rpm up to a maximum speed of 6000 rpm (corresponding to 4427 xg) in the popular 50ml and 15ml rotors The centrifuge lid features and automatic lid-lock that prevents it from being opened while the rotor is still in motion In addition a unique imbalance detection system is designed to automatically power down the motor in the event of improper loading of the rotor or any tube breakage Compact quiet and versatile are just a few of the features found in th