Learn More
Alkali Scientific Alkali Scientific™ Hermle Z207-H Compact Hematocrit Centrifuge
SDP
Catalog No. 503029571
Encompass_Preferred
Due to its compact design the Hermle Z207-H Hematocrit Centrifuge is the perfect centrifuge for even the smallest of lab spaces. It is equipped with a hematocrit rotor for 24 x capillaries. Accompanied by Hermle’s easy to use control panel , the unique EZ-Scroll™ touch pad, gives you ease of control and a modern feel. An advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf in increments of 10. In addition to retrieving saved programs, the easy to use front panel can be used to adjust accel/decel rates, run time, and change the end of run alarm tone. There is also a momentary spin function that will run while the button is pressed. Additionally the Hermle Z207-H features a sleep mode to save energy by shutting off after a certain time if not being used while on, which can be toggled on or off easily.
Catalog No. 50-302-9571 Supplier Alkali Scientific Supplier No. RS207H
Due to its compact design the Hermle Z207-H Hematocrit Centrifuge is the perfect centrifuge for even the smallest of lab spaces. It is equipped with a hematocrit rotor for 24 x capillaries. Accompanied by Hermle’s easy to use control panel , the unique EZ-Scroll™ touch pad, gives you ease of control and a modern feel. An advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf in increments of 10. In addition to retrieving saved programs, the easy to use front panel can be used to adjust accel/decel rates, run time, and change the end of run alarm tone. There is also a momentary spin function that will run while the button is pressed. Additionally the Hermle Z207-H features a sleep mode to save energy by shutting off after a certain time if not being used while on, which can be toggled on or off easily.