Learn More
Alkali Scientific Hermle Z216-M High Speed Micro
SDP
Catalog No. 503029640
Encompass_Preferred
Compact quiet and powerful are just a few of the features found on the Hermle Z216 series High Speed Microcentrifuge Add to this the versatility of eight available rotor options and the result is the ideal microcentrifuge for the modern molecular biology laboratory All common microtubes (0 2mL 0 5mL 1 5mL 2 0mL) can be accepted as well as PCR strips hematocrit tubes and even 5 0mL centrifuge tubes Advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf In addition to retrieving saved programs the easy to use front panel can adjust accel/decel rates and change the end of run alarm tone Despite a remarkably small footprint the Z216 series offers a uniquely designed 44 x 1 5mL dual row rotor nearly doubling the capacity when compared to traditional microcentrifuges 11 rotor options with capacity up to 44 x 1 5/2 0mL High speed 15000 rpm/21379 xg Accepts tubes from 0 2mL to 5mL Advanced controls with automatic rcf conversion
Catalog No. 50-302-9640 Supplier Alkali Scientific Supplier No. RS6202
Compact quiet and powerful are just a few of the features found on the Hermle Z216 series High Speed Microcentrifuge Add to this the versatility of eight available rotor options and the result is the ideal microcentrifuge for the modern molecular biology laboratory All common microtubes (0 2mL 0 5mL 1 5mL 2 0mL) can be accepted as well as PCR strips hematocrit tubes and even 5 0mL centrifuge tubes Advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf In addition to retrieving saved programs the easy to use front panel can adjust accel/decel rates and change the end of run alarm tone Despite a remarkably small footprint the Z216 series offers a uniquely designed 44 x 1 5mL dual row rotor nearly doubling the capacity when compared to traditional microcentrifuges 11 rotor options with capacity up to 44 x 1 5/2 0mL High speed 15000 rpm/21379 xg Accepts tubes from 0 2mL to 5mL Advanced controls with automatic rcf conversion