Learn More
Alkali Scientific Hermle Z216-MK Refrigerated Mi
SDP
Catalog No. 503029515
Encompass_Preferred
Compact quiet and powerful are just a few of the features found on the Hermle Z216-MK series Refrigerated Centrifuge Add to this the versatility of eight available rotor options and the result is the ideal microcentrifuge for the modern molecular biology laboratory All common microtubes (0 2ml 0 5ml 1 5ml 2 0ml) can be accepted as well as PCR strips hematocrit tubes and even 5 0ml centrifuge tubes Advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf In addition to retrieving saved programs the easy to use front panel can adjust accel/decel rates and change the end of run alarm tone Despite a remarkably small footprint the RS6203 series offers a uniquely designed 44 x 1 5ml dual row rotor nearly doubling the capacity when compared to traditional microcentrifuges In the Hermle Z216-MK model a powerful compressor driven cooling system maintains temperatures as low as -20 C and is designed to reach 4 C (from room temperature) in l
Catalog No. 50-302-9515 Supplier Alkali Scientific Supplier No. RS6203R
Add to Cart
Add to Cart
Request a Quote
Compact quiet and powerful are just a few of the features found on the Hermle Z216-MK series Refrigerated Centrifuge Add to this the versatility of eight available rotor options and the result is the ideal microcentrifuge for the modern molecular biology laboratory All common microtubes (0 2ml 0 5ml 1 5ml 2 0ml) can be accepted as well as PCR strips hematocrit tubes and even 5 0ml centrifuge tubes Advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf In addition to retrieving saved programs the easy to use front panel can adjust accel/decel rates and change the end of run alarm tone Despite a remarkably small footprint the RS6203 series offers a uniquely designed 44 x 1 5ml dual row rotor nearly doubling the capacity when compared to traditional microcentrifuges In the Hermle Z216-MK model a powerful compressor driven cooling system maintains temperatures as low as -20 C and is designed to reach 4 C (from room temperature) in l