Learn More
Alkali Scientific Hermle Z32 HK Super Speed Cent
SDP
Catalog No. 503029559
Encompass_Preferred
The Hermle Z32 HK Refrigerated Centirfuge utilizes much of the technology of our popular Z326 series with a larger and more powerful motor drive providing increased speed up to 20000 rpm (38007 xg) With 16 available rotor options the Z32 HK is not only ideal for high speed separations it would also be classified as a universal centrifuge offering economy angled and swinging bucket rotors for lower speed applications Tubes from 0 2ml up to 85ml can be accepted and rotors can quickly be exchanged without any required programming utilizing the instant rotor identification system Despite an impressive capacity of up to 4 x 145ml (or 6 x 85ml) the Hermle Z32-HK is surprisingly compact The refrigeration system is conveniently located behind the rotor chamber resulting in narrow design of less than 16 inches wide Advanced microprocessor controls allow for instant rotor identification which automatically converts the rotational speed to the relevant g-force (rcf) preventing the user from
Catalog No. 50-302-9559 Supplier Alkali Scientific Supplier No. RS6211
Add to Cart
Add to Cart
Request a Quote
The Hermle Z32 HK Refrigerated Centirfuge utilizes much of the technology of our popular Z326 series with a larger and more powerful motor drive providing increased speed up to 20000 rpm (38007 xg) With 16 available rotor options the Z32 HK is not only ideal for high speed separations it would also be classified as a universal centrifuge offering economy angled and swinging bucket rotors for lower speed applications Tubes from 0 2ml up to 85ml can be accepted and rotors can quickly be exchanged without any required programming utilizing the instant rotor identification system Despite an impressive capacity of up to 4 x 145ml (or 6 x 85ml) the Hermle Z32-HK is surprisingly compact The refrigeration system is conveniently located behind the rotor chamber resulting in narrow design of less than 16 inches wide Advanced microprocessor controls allow for instant rotor identification which automatically converts the rotational speed to the relevant g-force (rcf) preventing the user from