Learn More
Alkali Scientific Alkali Scientific™ Hermle Z327 Universal Centrifuges, 18000rpm
SDP
Catalog No. 503029598
Encompass_Preferred
The Herlme Z327 Series Universal lab centrifuge offer a wide assortment of rotor options (up to 19), making them ideal for a wide variety of research applications. Rotor exchange between swing out, micro, or high volume fixed angle rotors is very simple and can be done in a matter of seconds.  Swing out rotors accept up to 4 x 200 ml (16 x 15 ml or 8 x 50 ml) Swing out plate rotor accepts deepwell plates Extremely versatile, 17 rotor options Refrigerated model available (Z327-K) High speed rotors (up to 23,542 xg) Fixed angle rotors from 0.2 ml up to 85 ml
Catalog No. 50-302-9598 Supplier Alkali Scientific Supplier No. RS327
Add to Cart
Add to Cart
Request a Quote
The Herlme Z327 Series Universal lab centrifuge offer a wide assortment of rotor options (up to 19), making them ideal for a wide variety of research applications. Rotor exchange between swing out, micro, or high volume fixed angle rotors is very simple and can be done in a matter of seconds.  Swing out rotors accept up to 4 x 200 ml (16 x 15 ml or 8 x 50 ml) Swing out plate rotor accepts deepwell plates Extremely versatile, 17 rotor options Refrigerated model available (Z327-K) High speed rotors (up to 23,542 xg) Fixed angle rotors from 0.2 ml up to 85 ml