Learn More
Alkali Scientific Hermle Z36 HK Super Speed Cent
SDP
Catalog No. 503029590
Encompass_Preferred
The Hermle Z36 HK is the result of many years of experience in the design of universal centrifuges high speed centrifuges and compact microcentrifuges The result is a Super-Speed centrifuge with unmatched performance among benchtop centrifuges A robust motor drive powers a variety of high speed angled rotors to speeds that would generally only be achievable in large floor-standing ultra centirfuges 2 0ml microtube samples can reach forces above 65000xg while 50ml samples as high as 41410xg In addition to its high speed capabilities the Hermle Z36 HK doubles as a universal centrifuge with a 4 x 250ml swing out rotor (offering inserts for all common tube sizes) as well as a 2 x 3 position swing-out microplate rotor Advanced microprocessor controls allow for instant rotor identification which automatically converts the rotational speed to the relevant g-force (rcf) preventing the user from setting a speed above the capabilities of the installed rotor In addition to retrieving saved
Catalog No. 50-302-9590 Supplier Alkali Scientific Supplier No. RS036HK
Add to Cart
Add to Cart
Request a Quote
The Hermle Z36 HK is the result of many years of experience in the design of universal centrifuges high speed centrifuges and compact microcentrifuges The result is a Super-Speed centrifuge with unmatched performance among benchtop centrifuges A robust motor drive powers a variety of high speed angled rotors to speeds that would generally only be achievable in large floor-standing ultra centirfuges 2 0ml microtube samples can reach forces above 65000xg while 50ml samples as high as 41410xg In addition to its high speed capabilities the Hermle Z36 HK doubles as a universal centrifuge with a 4 x 250ml swing out rotor (offering inserts for all common tube sizes) as well as a 2 x 3 position swing-out microplate rotor Advanced microprocessor controls allow for instant rotor identification which automatically converts the rotational speed to the relevant g-force (rcf) preventing the user from setting a speed above the capabilities of the installed rotor In addition to retrieving saved