Learn More
Alkali Scientific Hermle Z496-K High Volume Floo
SDP
Catalog No. 503029669
Encompass_Preferred
The Hermle Z496-K centrifuges offer the highest tube/bottle capacity in our line Several rotors are available including a popular 4 position swinging bucket rotor with a wide offering of accessories for the centrifugation of tubes bottles plates and blood bags The high capacity rectangular buckets accept microplates directly or a variety of tube inserts for accomodating as many as 40 x 50ml conical or 96 x 15ml conical tubes per run Advanced microprocessor controls allow for instant rotor identificationwhich automatically converts the rotational speed to the relevant g-force (rcf) preventing the user from setting a speed above the capabilities of the installed rotor In addition to retrieving up to 99 saved programs the easy to use front panel can adjust accel/decel rates time and convert operation between rpm and rcf The powerful refrigeration system with a range of -20 to 40 C also features a precool function that ensures the temperature of the chamber is optimized for any appli
Catalog No. 50-302-9669 Supplier Alkali Scientific Supplier No. CP75A
Add to Cart
Add to Cart
Request a Quote
The Hermle Z496-K centrifuges offer the highest tube/bottle capacity in our line Several rotors are available including a popular 4 position swinging bucket rotor with a wide offering of accessories for the centrifugation of tubes bottles plates and blood bags The high capacity rectangular buckets accept microplates directly or a variety of tube inserts for accomodating as many as 40 x 50ml conical or 96 x 15ml conical tubes per run Advanced microprocessor controls allow for instant rotor identificationwhich automatically converts the rotational speed to the relevant g-force (rcf) preventing the user from setting a speed above the capabilities of the installed rotor In addition to retrieving up to 99 saved programs the easy to use front panel can adjust accel/decel rates time and convert operation between rpm and rcf The powerful refrigeration system with a range of -20 to 40 C also features a precool function that ensures the temperature of the chamber is optimized for any appli