Learn More
Alkali Scientific Alkali Scientific™ Hermle Z600-S3-TC SIEVA-3 Filtration Centrifuge with external temperature control, 120V
SDP
Catalog No. 503029633
Encompass_Preferred
The Hermle Z600-S3-TC SIEVA-3 centrifugal filtration system is the perfect centrifuge for cake filtration. The powerful Hermle Z600 SIEVA-3 reaches speeds up to 10,000rpm/7,825 xg, and features a transparent viewing port in the lid for close monitoring of samples. The stainless steel housing and parts makes the Hermle Z600-S3-TC SIEVA-3 corrosion resistant, and easy to clean. 500ml filtration baskets are available in both perorated and non perforated. Clarified liquids exit the unit via the drain located on the side of the unit. Advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf. In addition to retrieving saved programs, the easy to use front panel can adjust accel/decel rates, and change the end of run alarm tone.
Catalog No. 50-302-9633 Supplier Alkali Scientific Supplier No. RS9701
Add to Cart
Add to Cart
Request a Quote
The Hermle Z600-S3-TC SIEVA-3 centrifugal filtration system is the perfect centrifuge for cake filtration. The powerful Hermle Z600 SIEVA-3 reaches speeds up to 10,000rpm/7,825 xg, and features a transparent viewing port in the lid for close monitoring of samples. The stainless steel housing and parts makes the Hermle Z600-S3-TC SIEVA-3 corrosion resistant, and easy to clean. 500ml filtration baskets are available in both perorated and non perforated. Clarified liquids exit the unit via the drain located on the side of the unit. Advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf. In addition to retrieving saved programs, the easy to use front panel can adjust accel/decel rates, and change the end of run alarm tone.