Learn More
Medchemexpress LLC Herniarin-d3 (7-Methoxycoumarin-d3) | 1600535-74-7 | 179.19 | 1 MG
SDP
Catalog No. 5000414727
Encompass_Preferred
Herniarin-d3 is the deuterium labeled version of Herniarin, a natural coumarin found in some flowering plants known for its anticancer effects. It significantly decreases cell viability by inducing apoptosis in MCF-7 cells and also exhibits anti-dermatophytic activity. It can be used in the study of bladder cancer and breast cancer. Deuteration, the incorporation of stable heavy isotopes of hydrogen, is used in drug molecules largely as tracers for quantitation during drug development and has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potentially affects the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-14727 Supplier Medchemexpress LLC Supplier No. HYN1366S1MG
Herniarin-d3 is the deuterium labeled version of Herniarin, a natural coumarin found in some flowering plants known for its anticancer effects. It significantly decreases cell viability by inducing apoptosis in MCF-7 cells and also exhibits anti-dermatophytic activity. It can be used in the study of bladder cancer and breast cancer. Deuteration, the incorporation of stable heavy isotopes of hydrogen, is used in drug molecules largely as tracers for quantitation during drug development and has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potentially affects the pharmacokinetic and metabolic profiles of drugs