Learn More
Enzo Life Sciences HERP (human) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW97050100
Encompass_Preferred
Immunogen: Recombinant N-terminal cytoplasmic region of human HERP (aa 1-240). Species reactivity: Human. Applications: IP, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1697