Learn More
Bioss Herpes simplex virus 12
SDP
Catalog No. 5000066014
Encompass_Preferred
Herpes simplex virus 1 2 Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66014 Supplier Bioss Supplier No. BSM72225M
Herpes simplex virus 1 2 Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide