Learn More
Bioworld HERPES SIMPLEX VIRUS-8 MOSA
Supplier:  Bioworld 220601881
Encompass_Preferred
HERPES SIMPLEX VIRUS-8 MOSAI (RECOMBINANT): Recombinant Herpes Simplex Virus-8 Mosaic (HSV-8M) Source: E.Coli Description : E.coli derived recombinant. The protein contains the C-terminal immunodominant regions from ORF65 (140-170 a....
Catalog No. 50-253-288