Learn More
MyBioSource Herpes Simplex Virus GB 0.5mg ELISA Kit
SDP
Supplier:  MyBioSource MBS5680240.5 MG
Encompass_Preferred
The MyBioSource Herpes Simplex Virus GB ELISA Kit quantitatively measures HSV GB in various biological fluids with a 0.156-10 ng/mL range. It includes materials for 96 tests using a sandwich ELISA format with specific antibodies, biotinylated detection antibody, HRP conjugate, and TMB substrate for colorimetric detection at 450nm, ensuring high sensitivity.
Catalog No. NC2032988