Learn More
Diagnostic Biosystems HERPES SIMPLEX VIRUS II HSV II Herpes Simplex Virus II (HSV II)
Catalog No. 502407502
Encompass_Preferred
This antibody reacts with herpes simplex virus type 2. It is specific for the viral glycoprotein D (gD) protein.
Catalog No. 50-240-7502 Supplier Diagnostic Biosystems Supplier No. PDM542
This antibody reacts with herpes simplex virus type 2. It is specific for the viral glycoprotein D (gD) protein.