Learn More
AdipoGen Herquline A (250 µg)
SDP
Supplier:  AdipoGen AGCN20170C250
Encompass_Preferred
Chemical. CAS: 71812-08-3. Formula: C19H26N2O2. MW: 314.4. Isolated from Penicillium herquei. Weak blood platelet aggregation inhibitor compared to Herquline B.
Catalog No. 50-205-3562