Learn More
Creative Biomart Recombinant HSV-8 ORF65 and ORF8 Antigen, GST-tagged
Catalog No. 502084713
Encompass_Preferred
The E. coli derived recombinant protein contains the C-terminal immunodominant regions from ORF65 140-170 a.a. and N-terminal regions from ORF8 32-62 a.a. The protein is fused with a GST tag.
Catalog No. 50-208-4713 Supplier Creative Biomart Supplier No. HER335
The E. coli derived recombinant protein contains the C-terminal immunodominant regions from ORF65 140-170 a.a. and N-terminal regions from ORF8 32-62 a.a. The protein is fused with a GST tag.