Learn More
Aviva Systems Biology Corp HES1 Antibody - N-terminal region (ARP32372_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32372_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HES1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HES1. Alternate names: HHL, HRY, HES-1, bHLHb39ApplicationIF, IP, WB, IHC. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54550