Learn More
ABclonal Technology HES1 Rabbit pAb
SDP
Catalog No. 5000116054
Encompass_Preferred
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.
Catalog No. 50-001-16054 Supplier ABclonal Technology Supplier No. A1170520UL
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.