Learn More
Aviva Systems Biology Corp HES4 Antibody - middle region (ARP36000_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36000_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HES4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HES4. Alternate names: bHLHb42ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-55962