Learn More
Aviva Systems Biology Corp HES4 Antibody - N-terminal region (ARP35999_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35999_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HES4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HES4. Alternate names: bHLHb42ApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rat
Catalog No. 50-014-55961