Learn More
Aviva Systems Biology Corp HES5 Antibody - middle region (ARP50367_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50367_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HES5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HES5. Alternate names: bHLHb38ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-63013