Learn More
Aviva Systems Biology Corp Hes7 Antibody - middle region (ARP37926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hes7. It was validated on Western Blot. Hes7,hairy and enhancer of split 7 (Drosophila),bHLHb37; Hes7,transcription factor HES-7,transcription factor HES-7,mHes7,bHLH factor Hes7,OTTMUSP00000006176The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: bHLHb37ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-703