Learn More
Thermo Scientific Heater Thermostat 1Pc
Supplier: Thermo Scientific 7000560157
Thermocouple and tilt switch kit for patio heaters contains 304 stainless steel thermocouple sensor to detect pilot flame and activate gas valve, and ABS plastic tilt switch to automatically shut off gas if heater tips over, designed as replacement parts to repair malfunctioning or damaged thermocouple and tilt sensors in propane fired radiant patio heaters.