Learn More
Apexbio Technology LLC Cell-permeable, reversible, and ATP-competitive Aurora B kinase inhibitor 422513-13-1. MFCD18074526
SDP
Supplier:  Apexbio Technology LLC A41185
Encompass_Preferred
Cell-permeable, reversible, and ATP-competitive Aurora B kinase inhibitor 422513-13-1. MFCD18074526
Catalog No. 50-101-4180