Learn More
Medchemexpress LLC HY-12054 5mg Medchemexpress, Hesperadin CAS:422513-13-1 Purity:>98%
SDP
Catalog No. 502028847
Encompass_Preferred
Medchemexpress, HY-12054 5mg Hesperadin CAS:422513-13-1 Hesperadin is an ATP competitive indolinone inhibitor of Aurora A and B. Hesperadin inhibits Aurora B with an IC50 of 250 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8847 Supplier Medchemexpress LLC Supplier No. HY120545MG
Medchemexpress, HY-12054 5mg Hesperadin CAS:422513-13-1 Hesperadin is an ATP competitive indolinone inhibitor of Aurora A and B. Hesperadin inhibits Aurora B with an IC50 of 250 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.