Learn More
Cayman Chemical Hesperetin-13C-d3, Purity: ≥99% deuterated forms (d1-d3)., CAS: 2750534-85-9, 1 mg
SDP
Catalog No. 502940181
Encompass_Preferred
An internal standard for the quantification of hesperetin by GC- or LC-MS
Catalog No. 50-294-0181 Supplier Cayman Chemical Supplier No. 288061 MG
An internal standard for the quantification of hesperetin by GC- or LC-MS