Learn More
Cayman Chemical HesperetIn-13C-d3 500ug
SDP
Catalog No. 502941150
Encompass_Preferred
An internal standard for the quantification of hesperetin by GC- or LC-MS
Catalog No. 50-294-1150 Supplier Cayman Chemical Supplier No. 28806500 UG
An internal standard for the quantification of hesperetin by GC- or LC-MS