Learn More
Medchemexpress LLC HY-N0168 50mg Medchemexpress, Hesperetin CAS:520-33-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN016850MG
Encompass_Preferred
Medchemexpress, HY-N0168 50mg Hesperetin CAS:520-33-2 Hesperetin is a natural flavanone, and acts as a potent and broad-spectrum inhibitor against human UGT activity. Hesperetin induces apoptosis via p38 MAPK activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3441