Learn More
Medchemexpress LLC Hesperetin | 520-33-2 | 99.1% | 302.28 | 1 ML
SDP
Catalog No. 5000427403
Encompass_Preferred
Hesperetin is a natural flavanone found in citrus, acting as a potent and orally active broad-spectrum inhibitor against human UGT activity. It induces apoptosis via p38 MAPK activation and demonstrates a range of bioactivities, including antioxidant, anti-inflammatory, and anti-cancer properties. Hesperetin can also induce cell-cycle arrest at the G2/M phase, reduce Bcl-2, enhance BaxM, and induce apoptosis by inhibiting the NF-κB receptor.
  • Natural flavanone from citrus
  • Potent and orally active broad-spectrum inhibitor against human UGT activity
  • Induces apoptosis via p38 MAPK activation
  • Exhibits antioxidant, anti-inflammatory, and anti-cancer properties
  • Induces cell-cycle arrest at the G2/M phase
  • Reduces Bcl-2 and enhances BaxM
  • Inhibits the NF-κB receptor
Catalog No. 50-004-27403 Supplier Medchemexpress LLC Supplier No. HYN016810MM/1ML
Hesperetin is a natural flavanone found in citrus, acting as a potent and orally active broad-spectrum inhibitor against human UGT activity. It induces apoptosis via p38 MAPK activation and demonstrates a range of bioactivities, including antioxidant, anti-inflammatory, and anti-cancer properties. Hesperetin can also induce cell-cycle arrest at the G2/M phase, reduce Bcl-2, enhance BaxM, and induce apoptosis by inhibiting the NF-κB receptor.
  • Natural flavanone from citrus
  • Potent and orally active broad-spectrum inhibitor against human UGT activity
  • Induces apoptosis via p38 MAPK activation
  • Exhibits antioxidant, anti-inflammatory, and anti-cancer properties
  • Induces cell-cycle arrest at the G2/M phase
  • Reduces Bcl-2 and enhances BaxM
  • Inhibits the NF-κB receptor