Learn More
Cayman Chemical Hesperetin 7-O-glucoside, Purity: ≥98%, CAS: 31712-49-9, 25 mg
SDP
Catalog No. 502950155
Encompass_Preferred
A flavanone glucoside; active against H. pylori at 0.5 mM; scavenges DPPH radicals (IC50 = 2.04 mM) and inhibits the activity of intestinal maltase and HMG-CoA reductase (Kis = 1,800 and 9.8 µM, respectively, for the human enzymes) in cell-free assays; prevents decreases in bone mineral density in the femur of ovariectomized rats at 2.5 and 5 g/kg diet
Catalog No. 50-295-0155 Supplier Cayman Chemical Supplier No. 3454525 MG
A flavanone glucoside; active against H. pylori at 0.5 mM; scavenges DPPH radicals (IC50 = 2.04 mM) and inhibits the activity of intestinal maltase and HMG-CoA reductase (Kis = 1,800 and 9.8 µM, respectively, for the human enzymes) in cell-free assays; prevents decreases in bone mineral density in the femur of ovariectomized rats at 2.5 and 5 g/kg diet